Skip to Content
Merck

BDPH9BCCE1EC

Bide Pharmatech

Methyl 2-(2-(2-aminoethoxy)ethoxy)acetate hydrochloride

Synonym(s):

methyl 2-[2-(2-aminoethoxy)ethoxy]acetate hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H16O4N1Cl1
CAS Number:
Molecular Weight:
213.66
MDL number:
Product BDPH9BCCE1EC is not currently sold in your country.


Inchi Code

1S/C7H15NO4.ClH/c1-10-7(9)6-12-5-4-11-3-2-8;/h2-6,8H2,1H3;1H

Inchi Key

LJGPEQCMODSTPF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to White to Yellow Solid or Semi-Solid or Liquid or Lump



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library