Inchi Code
1S/C15H16N2O2S/c16-13-7-9-14(10-8-13)20(18,19)17-11-3-5-12-4-1-2-6-15(12)17/h1-2,4,6-10H,3,5,11,16H2
Inchi Key
OFHUMRFYQYBKIN-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 4-((3,4-Dihydroquinolin-1(2H)-yl)sulfonyl)aniline
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
