Skip to Content
Merck

BDPH9BCCE80A

Bide Pharmatech

1-Methyl-4-(piperidin-3-ylmethyl)piperazine trihydrochloride

Synonym(s):

1-methyl-4-(3-piperidinylmethyl)piperazine trihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H26N3Cl3
Molecular Weight:
306.71
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H23N3.3ClH/c1-13-5-7-14(8-6-13)10-11-3-2-4-12-9-11;;;/h11-12H,2-10H2,1H3;3*1H

Inchi Key

HTMDHBRVDDGQSC-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library