Skip to Content
Merck

BDPH9BCCEB06

Bide Pharmatech

3-Bromo-5-(1-hydroxyethyl)pyridine

Synonym(s):

1-(5-bromo-3-pyridinyl)ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H8O1N1Br1
CAS Number:
Molecular Weight:
202.05
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H8BrNO/c1-5(10)6-2-7(8)4-9-3-6/h2-5,10H,1H3

Inchi Key

SRELUKWTQNFSRR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 3-Bromo-5-(1-hydroxyethyl)pyridine


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library