Skip to Content
Merck

BDPH9BCCEDF4

Bide Pharmatech

2-(2-Iodophenyl)-N,N-dimethylacetamide

Synonym(s):

2-(2-iodophenyl)-N,N-dimethylacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12O1N1I1
CAS Number:
Molecular Weight:
289.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H12INO/c1-12(2)10(13)7-8-5-3-4-6-9(8)11/h3-6H,7H2,1-2H3

Inchi Key

BPDRJYYNQKBYBG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library