Skip to Content
Merck

BDPH9BCCEF3A

Bide Pharmatech

N,3-Dimethylbutan-2-amine

Synonym(s):

N,3-dimethyl-2-butanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H15N1
CAS Number:
Molecular Weight:
101.19
MDL number:
Product BDPH9BCCEF3A is not currently sold in your country.


Inchi Code

1S/C6H15N/c1-5(2)6(3)7-4/h5-7H,1-4H3

Inchi Key

LJLWVVCWBURGCC-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under N,3-Dimethylbutan-2-amine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library