Skip to Content
Merck

BDPH9BCCFAC5

Bide Pharmatech

1-(5-Bromo-1H-pyrrol-3-yl)-N-methylmethanamine

Synonym(s):

N-[(5-bromo-1H-pyrrol-3-yl)methyl]-N-methylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H9N2Br1
CAS Number:
Molecular Weight:
189.05
MDL number:
Product BDPH9BCCFAC5 is not currently sold in your country.


Inchi Code

1S/C6H9BrN2/c1-8-3-5-2-6(7)9-4-5/h2,4,8-9H,3H2,1H3

Inchi Key

ITOMXMSCUOKHLN-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

Liquid or solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library