Skip to Content
Merck

BDPH9BCCFB4F

Bide Pharmatech

6-Chloroquinoxaline-2,3-diol

Synonym(s):

6-chloro-2,3-quinoxalinediol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H5O2N2Cl1
CAS Number:
Molecular Weight:
196.59
MDL number:
Product BDPH9BCCFB4F is not currently sold in your country.


Inchi Code

1S/C8H5ClN2O2/c9-4-1-2-5-6(3-4)11-8(13)7(12)10-5/h1-3H,(H,10,12)(H,11,13)

Inchi Key

RNOLFZACEWWIHP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 6-Chloroquinoxaline-2,3-diol


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library