Skip to Content
Merck

BDPH9BCCFC73

Bide Pharmatech

5-Bromo-2-fluoro-1-methoxy-3-methylbenzene

Synonym(s):

5-bromo-2-fluoro-3-methylphenyl methyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H8O1Br1F1
CAS Number:
Molecular Weight:
219.05
MDL number:
Product BDPH9BCCFC73 is not currently sold in your country.


Inchi Code

1S/C8H8BrFO/c1-5-3-6(9)4-7(11-2)8(5)10/h3-4H,1-2H3

Inchi Key

BNLXSNQTRFKATL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library