Skip to Content
Merck

BDPH9BCCFF6E

Bide Pharmatech

4,4-Difluoro-1-methylpyrrolidin-3-ol

Synonym(s):

4,4-difluoro-1-methylpyrrolidin-3-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H9O1N1F2
CAS Number:
Molecular Weight:
137.13
MDL number:
Product BDPH9BCCFF6E is not currently sold in your country.


Inchi Code

1S/C5H9F2NO/c1-8-2-4(9)5(6,7)3-8/h4,9H,2-3H2,1H3

Inchi Key

TYYKAQBZBXSNBA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid or powder



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library