Skip to Content
Merck

BDPH9BCD0601

Bide Pharmatech

4-Bromo-6-chloro-2-methylpyridin-3-amine

Synonym(s):

4-bromo-6-chloro-2-methylpyridin-3-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6N2Cl1Br1
CAS Number:
Molecular Weight:
221.48
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6BrClN2/c1-3-6(9)4(7)2-5(8)10-3/h2H,9H2,1H3

Inchi Key

XFRUXXUMLANNIE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library