Skip to Content

Legal compliance reminder: purchasers must obtain all necessary licenses and comply with applicable laws (see General Terms & Conditions of Sale). We reserve the right to request additional information.

Merck

Skip To

BDPH9BCD0965

Bide Pharmatech

4,6-Difluoro-1H-indole-2,3-dione

Synonym(s):

4,6-difluoro-1H-indole-2,3-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C8H3O2N1F2
CAS Number:
Molecular Weight:
183.11
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H3F2NO2/c9-3-1-4(10)6-5(2-3)11-8(13)7(6)12/h1-2H,(H,11,12,13)

Inchi Key

YIVXDNUTNIKQMY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid



Still not finding the right product?

Explore all of our products under 4,6-Difluoro-1H-indole-2,3-dione


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library