Inchi Code
1S/C7H15N.ClH/c1-2-7(8)6-4-3-5-6;/h6-7H,2-5,8H2,1H3;1H
Inchi Key
QLVATDBARBKCCH-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 1-Cyclobutylpropan-1-amine hydrochloride
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
