Skip to Content
Merck

BDPH9BCD0BB1

Bide Pharmatech

5-Chlorothiazole-4-carboxylic acid

Synonym(s):

5-chlorothiazole-4-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H2O2N1Cl1S1
CAS Number:
Molecular Weight:
163.58
MDL number:
Product BDPH9BCD0BB1 is not currently sold in your country.


Inchi Code

1S/C4H2ClNO2S/c5-3-2(4(7)8)6-1-9-3/h1H,(H,7,8)

Inchi Key

VFCFYIDCFCQUFD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 5-Chlorothiazole-4-carboxylic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library