Skip to Content

BDPH9BCD0BD8

Bide Pharmatech

2-(Oxetan-3-yloxy)aniline

Synonym(s):

2-(3-oxetanyloxy)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O2N1
CAS Number:
Molecular Weight:
165.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11NO2/c10-8-3-1-2-4-9(8)12-7-5-11-6-7/h1-4,7H,5-6,10H2

Inchi Key

ZYDPAUQAFXSHAB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 2-(Oxetan-3-yloxy)aniline



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library