Skip to Content
Merck

BDPH9BCD0D4B

Bide Pharmatech

4-Methoxy-2,3,5-trimethylbenzaldehyde

Synonym(s):

4-methoxy-2,3,5-trimethylbenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H14O2
CAS Number:
Molecular Weight:
178.23
MDL number:
Product BDPH9BCD0D4B is not currently sold in your country.


Inchi Code

1S/C11H14O2/c1-7-5-10(6-12)8(2)9(3)11(7)13-4/h5-6H,1-4H3

Inchi Key

RMFKBXSYAGJVQR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid or solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library