Skip to Content
Merck

BDPH9BCD102B

Bide Pharmatech

2-Phenylpyrimidin-5-amine

Synonym(s):

2-phenyl-5-pyrimidinamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9N3
CAS Number:
Molecular Weight:
171.2
MDL number:
Product BDPH9BCD102B is not currently sold in your country.


Inchi Code

1S/C10H9N3/c11-9-6-12-10(13-7-9)8-4-2-1-3-5-8/h1-7H,11H2

Inchi Key

YUPYDSQWLFBBJJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to Pale-yellow Powder or Crystals



Still not finding the right product?

Explore all of our products under 2-Phenylpyrimidin-5-amine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library