Skip to Content
Merck

BDPH9BCD1CCD

Bide Pharmatech

4-Chloro-6-methoxy-2,3-dihydro-1H-inden-1-one

Synonym(s):

4-chloro-6-methoxy-1-indanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O2Cl1
CAS Number:
Molecular Weight:
196.63
MDL number:
Product BDPH9BCD1CCD is not currently sold in your country.


Inchi Code

1S/C10H9ClO2/c1-13-6-4-8-7(9(11)5-6)2-3-10(8)12/h4-5H,2-3H2,1H3

Inchi Key

AXWJUEOGJHUJMW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library