Skip to Content
Merck

Skip To

BDPH9BCD20C2

Bide Pharmatech

2-Amino-3-(thiazol-5-yl)propanoic acid hydrochloride

Synonym(s):

2-amino-3-(thiazol-5-yl)propanoic acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H9O2N2Cl1S1
CAS Number:
Molecular Weight:
208.67
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Purity

97%

Inchi Code

1S/C6H8N2O2S.ClH/c7-5(6(9)10)1-4-2-8-3-11-4;/h2-3,5H,1,7H2,(H,9,10);1H

Inchi Key

WPTRJRFKLLPZND-UHFFFAOYSA-N

Shipping Temperature

Normal

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library