Skip to Content
Merck

BDPH9BCD22EB

Bide Pharmatech

1-((4-Methyl-4H-1,2,4-triazol-3-yl)thio)butan-2-amine

Synonym(s):

1-[(4-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]-2-butanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H14N4S1
CAS Number:
Molecular Weight:
186.28
MDL number:
Product BDPH9BCD22EB is not currently sold in your country.


Inchi Code

1S/C7H14N4S/c1-3-6(8)4-12-7-10-9-5-11(7)2/h5-6H,3-4,8H2,1-2H3

Inchi Key

LFNUDWAXBARNQH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library