Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C12H14BNO2/c14-9-12(7-1-2-8-12)10-3-5-11(6-4-10)13(15)16/h3-6,15-16H,1-2,7-8H2
Inchi Key
ZNINYILPLNZOEJ-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under [4-(1-Cyanocyclopentyl)phenyl]boronic acid
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
