Skip to Content
Merck

BDPH9BCD2CF1

Bide Pharmatech

(5-Chloropyridin-2-yl)methanol

Synonym(s):

(5-chloro-2-pyridinyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6O1N1Cl1
CAS Number:
Molecular Weight:
143.57
MDL number:
Product BDPH9BCD2CF1 is not currently sold in your country.


Inchi Code

1S/C6H6ClNO/c7-5-1-2-6(4-9)8-3-5/h1-3,9H,4H2

Inchi Key

JJNKKECPOPWYNY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid



Still not finding the right product?

Explore all of our products under (5-Chloropyridin-2-yl)methanol


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library