Skip to Content
Merck

BDPH9BCD2DE2

Bide Pharmatech

Dibenzo[b,d]furan-1-ylboronic acid

Synonym(s):

dibenzo[b,d]furan-1-ylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H9O3B1
CAS Number:
Molecular Weight:
212.01
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H9BO3/c14-13(15)9-5-3-7-11-12(9)8-4-1-2-6-10(8)16-11/h1-7,14-15H

Inchi Key

JOFBTOVNZIUWPX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to pale yellow powder or crystal



Still not finding the right product?

Explore all of our products under Dibenzo[b,d]furan-1-ylboronic acid


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library