Skip to Content

BDPH9BCD320D

Bide Pharmatech

Tris(4-(phenylethynyl)phenyl)amine

Synonym(s):

tris(4-(phenylethynyl)phenyl)amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C42H27N1
CAS Number:
Molecular Weight:
545.68
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C42H27N/c1-4-10-34(11-5-1)16-19-37-22-28-40(29-23-37)43(41-30-24-38(25-31-41)20-17-35-12-6-2-7-13-35)42-32-26-39(27-33-42)21-18-36-14-8-3-9-15-36/h1-15,22-33H

Inchi Key

RWWNAZWAWXEDKN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under Tris(4-(phenylethynyl)phenyl)amine


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library