Skip to Content
Merck

BDPH9BCD3255

Bide Pharmatech

5-(4-Bromophenyl)pyrimidine

Synonym(s):

5-(4-bromophenyl)pyrimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H7N2Br1
CAS Number:
Molecular Weight:
235.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H7BrN2/c11-10-3-1-8(2-4-10)9-5-12-7-13-6-9/h1-7H

Inchi Key

GJBAOBLGEYEVMJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 5-(4-Bromophenyl)pyrimidine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library