Skip to Content
Merck

BDPH9BCD3322

Bide Pharmatech

3-(Benzyloxy)azetidine hydrochloride

Synonym(s):

3-(benzyloxy)azetidine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H14O1N1Cl1
Molecular Weight:
199.68
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H13NO.ClH/c1-2-4-9(5-3-1)8-12-10-6-11-7-10;/h1-5,10-11H,6-8H2;1H

Inchi Key

BQTSSUSLZRYTQB-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library