Skip to Content
Merck

BDPH9BCD3371

Bide Pharmatech

N2-(4-Aminophenyl)-N4,N4,6-trimethylpyrimidine-2,4-diamine

Synonym(s):

N~2~-(4-aminophenyl)-N~4~,N~4~,6-trimethyl-2,4-pyrimidinediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H17N5
CAS Number:
Molecular Weight:
243.31
MDL number:
Product BDPH9BCD3371 is not currently sold in your country.


Inchi Code

1S/C13H17N5/c1-9-8-12(18(2)3)17-13(15-9)16-11-6-4-10(14)5-7-11/h4-8H,14H2,1-3H3,(H,15,16,17)

Inchi Key

LYQRHCXHOCPMFZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library