Skip to Content
Merck

Skip To

BDPH9BCD3EA2

Bide Pharmatech

Ethyl 5-hydroxy-1H-pyrazole-3-carboxylate

Synonym(s):

ethyl 5-hydroxy-1H-pyrazole-3-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H8O3N2
CAS Number:
Molecular Weight:
156.14
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H8N2O3/c1-2-11-6(10)4-3-5(9)8-7-4/h3H,2H2,1H3,(H2,7,8,9)

Inchi Key

FGCPAXRNQIOISG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library