Skip to Content
Merck

BDPH9BCD3F76

Bide Pharmatech

Bis(4-(tert-butyl)phenyl)amine

Synonym(s):

4-tert-butyl-N-(4-tert-butylphenyl)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H27N1
CAS Number:
Molecular Weight:
281.44
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H27N/c1-19(2,3)15-7-11-17(12-8-15)21-18-13-9-16(10-14-18)20(4,5)6/h7-14,21H,1-6H3

Inchi Key

OPEKHRGERHDLRK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

90%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Slightly pale yellow



Still not finding the right product?

Explore all of our products under Bis(4-(tert-butyl)phenyl)amine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark

hstatements



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library