Skip to Content
Merck

BDPH9BCD4850

Bide Pharmatech

1-(3-(Trifluoromethoxy)phenyl)propan-1-amine hydrochloride

Synonym(s):

1-(3-(trifluoromethoxy)phenyl)propan-1-amine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H13O1N1Cl1F3
CAS Number:
Molecular Weight:
255.67
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H12F3NO.ClH/c1-2-9(14)7-4-3-5-8(6-7)15-10(11,12)13;/h3-6,9H,2,14H2,1H3;1H

Inchi Key

FYDVWLSWTITCCB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library