Skip to Content
Merck

BDPH9BCD496E

Bide Pharmatech

tert-Butyl 4-(3-hydroxyphenyl)piperazine-1-carboxylate

Synonym(s):

tert-butyl 4-(3-hydroxyphenyl)-1-piperazinecarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H22O3N2
CAS Number:
Molecular Weight:
278.35
MDL number:
Product BDPH9BCD496E is not currently sold in your country.


Country of Origin

CN

Physical Form

Yellow to Brown Solid

Inchi Code

1S/C15H22N2O3/c1-15(2,3)20-14(19)17-9-7-16(8-10-17)12-5-4-6-13(18)11-12/h4-6,11,18H,7-10H2,1-3H3

Inchi Key

MIQHJGPQPMQVOT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library