Skip to Content
Merck

BDPH9BCD5419

Bide Pharmatech

6-Formylpicolinic acid

Synonym(s):

6-formyl-2-pyridinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5O3N1
Molecular Weight:
151.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H5NO3/c9-4-5-2-1-3-6(8-5)7(10)11/h1-4H,(H,10,11)

Inchi Key

DAUQNAWDINBWER-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library