Skip to Content
Merck

BDPH9BCD561B

Bide Pharmatech

Methyl 3-(piperidin-4-yl)benzoate hydrochloride

Synonym(s):

methyl 3-(4-piperidinyl)benzoate hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H18O2N1Cl1
CAS Number:
Molecular Weight:
255.74
MDL number:
Product BDPH9BCD561B is not currently sold in your country.


Inchi Code

1S/C13H17NO2.ClH/c1-16-13(15)12-4-2-3-11(9-12)10-5-7-14-8-6-10;/h2-4,9-10,14H,5-8H2,1H3;1H

Inchi Key

CTBDUSKBEWVQOC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library