Skip to Content

BDPH9BCD624E

Bide Pharmatech

4-(Chloromethyl)-2,6-dimethylpyridine hydrochloride

Synonym(s):

4-(chloromethyl)-2,6-dimethylpyridine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H11N1Cl2
CAS Number:
Molecular Weight:
192.09
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H10ClN.ClH/c1-6-3-8(5-9)4-7(2)10-6;/h3-4H,5H2,1-2H3;1H

Inchi Key

WPUGKCDNYQMSHO-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

97%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Inert atmosphere, store in freezer, under -20°C



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library