Skip to Content
Merck

BDPH9BCD659E

Bide Pharmatech

3,5-Dichloro-N-(3-chloro-4-(4-chlorophenoxy)phenyl)-2-hydroxybenzamide

Synonym(s):

3,5-dichloro-N-(3-chloro-4-(4-chlorophenoxy)phenyl)-2-hydroxybenzimidic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product BDPH9BCD659E is not currently sold in your country.


Inchi Code

1S/C19H11Cl4NO3/c20-10-1-4-13(5-2-10)27-17-6-3-12(9-15(17)22)24-19(26)14-7-11(21)8-16(23)18(14)25/h1-9,25H,(H,24,26)

Inchi Key

HPIKHGXRCDJEKT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Off-white to light yellow solid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library