Skip to Content
Merck

BDPH9BCD6EF7

Bide Pharmatech

2-Methyl-2-azaspiro[3.3]heptan-6-amine dihydrochloride

Synonym(s):

2-methyl-2-azaspiro[3.3]heptan-6-amine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H16N2Cl2
CAS Number:
Molecular Weight:
199.12
MDL number:
Product BDPH9BCD6EF7 is not currently sold in your country.


Inchi Code

1S/C7H14N2.2ClH/c1-9-4-7(5-9)2-6(8)3-7;;/h6H,2-5,8H2,1H3;2*1H

Inchi Key

WAFDVVXCCJNCED-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library