Skip to Content
Merck

BDPH9BCD72A4

Bide Pharmatech

6-Bromo-5,8-difluoroquinoline

Synonym(s):

6-bromo-5,8-difluoroquinoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H4N1Br1F2
CAS Number:
Molecular Weight:
244.04
MDL number:
Product BDPH9BCD72A4 is not currently sold in your country.


Inchi Code

1S/C9H4BrF2N/c10-6-4-7(11)9-5(8(6)12)2-1-3-13-9/h1-4H

Inchi Key

GVFUBMHGLPCFNH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Powder or crystals

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?

Explore all of our products under 6-Bromo-5,8-difluoroquinoline



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library