Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C10H8O4/c1-2-6-3-7(9(11)12)5-8(4-6)10(13)14/h2-5H,1H2,(H,11,12)(H,13,14)
Inchi Key
RFRNGRYAJCOIAN-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 5-Vinylisophthalic acid
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
