Skip to Content
Merck

BDPH9BCD7BE8

Bide Pharmatech

1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine

Synonym(s):

1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-ylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H6N3F3
CAS Number:
Molecular Weight:
165.12
MDL number:
Product BDPH9BCD7BE8 is not currently sold in your country.


Inchi Code

1S/C5H6F3N3/c1-11-2-3(9)4(10-11)5(6,7)8/h2H,9H2,1H3

Inchi Key

OHASPUNAEXVWEP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Keep in dark place, inert atmosphere, 2-8°C



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library