Skip to Content
Merck

BDPH9BCD8380

Bide Pharmatech

3-Methyl-2-oxopiperidine-3-carboxylic acid

Synonym(s):

3-methyl-2-oxo-3-piperidinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H11O3N1
CAS Number:
Molecular Weight:
157.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H11NO3/c1-7(6(10)11)3-2-4-8-5(7)9/h2-4H2,1H3,(H,8,9)(H,10,11)

Inchi Key

ZHFYGKPIJJBFJH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder to crystal



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library