Skip to Content
Merck

BDPH9BCD85A3

Bide Pharmatech

tert-Butyl 3-((4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl)azetidine-1-carboxylate

Synonym(s):

tert-butyl 3-((4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl)azetidine-1-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H28O4N1B1
CAS Number:
Molecular Weight:
297.2
MDL number:
Product BDPH9BCD85A3 is not currently sold in your country.


Inchi Code

1S/C15H28BNO4/c1-13(2,3)19-12(18)17-9-11(10-17)8-16-20-14(4,5)15(6,7)21-16/h11H,8-10H2,1-7H3

Inchi Key

ZBUVYHVQMDQQLA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library