Skip to Content
Merck

BDPH9BCD8D3A

Bide Pharmatech

6-Methoxyquinazolin-4-ol

Synonym(s):

6-methoxy-4(3H)-quinazolinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H8O2N2
CAS Number:
Molecular Weight:
176.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H8N2O2/c1-13-6-2-3-8-7(4-6)9(12)11-5-10-8/h2-5H,1H3,(H,10,11,12)

Inchi Key

NOFVNLZQAOGUIT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 6-Methoxyquinazolin-4-ol


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library