Skip to Content
Merck

BDPH9BCD92EE

Bide Pharmatech

1-(Cyclobutylmethyl)-1H-imidazole-4-carboxylic acid

Synonym(s):

1-(cyclobutylmethyl)-1H-imidazole-4-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H12O2N2
CAS Number:
Molecular Weight:
180.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H12N2O2/c12-9(13)8-5-11(6-10-8)4-7-2-1-3-7/h5-7H,1-4H2,(H,12,13)

Inchi Key

PQFSBOOTPMIKFP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library