Skip to Content
Merck

BDPH9BCD995D

Bide Pharmatech

Azetidine-1-carbonyl chloride

Synonym(s):

azetidine-1-carbonyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H6O1N1Cl1
CAS Number:
Molecular Weight:
119.55
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H6ClNO/c5-4(7)6-2-1-3-6/h1-3H2

Inchi Key

LJPIQCGBOGKQOK-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under Azetidine-1-carbonyl chloride



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library