Skip to Content
Merck

BDPH9BCDA7DE

Bide Pharmatech

Methyl 3-hydroxy-2-methylenepentanoate

Synonym(s):

methyl 3-hydroxy-2-methylenepentanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H12O3
CAS Number:
Molecular Weight:
144.17
MDL number:
Product BDPH9BCDA7DE is not currently sold in your country.


Inchi Code

1S/C7H12O3/c1-4-6(8)5(2)7(9)10-3/h6,8H,2,4H2,1,3H3

Inchi Key

WXZUVXFBEAOGOZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library