Skip to Content
Merck

BDPH9BCDABC3

Bide Pharmatech

1-Bromo-3-methylbutan-2-one

Synonym(s):

1-bromo-3-methyl-2-butanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H9O1Br1
CAS Number:
Molecular Weight:
165.03
MDL number:
Product BDPH9BCDABC3 is not currently sold in your country.


Inchi Code

1S/C5H9BrO/c1-4(2)5(7)3-6/h4H,3H2,1-2H3

Inchi Key

NNTPEAXKKUPBHQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid



Still not finding the right product?

Explore all of our products under 1-Bromo-3-methylbutan-2-one



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library