Skip to Content
Merck

BDPH9BCDB7C4

Bide Pharmatech

4-Phenyl-2-(pyridin-4-yl)thiazole-5-carboxylic acid

Synonym(s):

4-phenyl-2-(4-pyridinyl)-1,3-thiazole-5-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H10O2N2S1
Molecular Weight:
282.32
MDL number:
Product BDPH9BCDB7C4 is not currently sold in your country.


Inchi Code

1S/C15H10N2O2S/c18-15(19)13-12(10-4-2-1-3-5-10)17-14(20-13)11-6-8-16-9-7-11/h1-9H,(H,18,19)

Inchi Key

XIUFXYLUJWNFRF-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library