Skip to Content
Merck

BDPH9BCDBA3C

Bide Pharmatech

2-(Difluoromethoxy)ethan-1-amine

Synonym(s):

2-(difluoromethoxy)ethan-1-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C3H7O1N1F2
CAS Number:
Molecular Weight:
111.09
MDL number:
Product BDPH9BCDBA3C is not currently sold in your country.


Inchi Code

1S/C3H7F2NO/c4-3(5)7-2-1-6/h3H,1-2,6H2

Inchi Key

XVHQHDWOVYAABP-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Colorless to ligh yellow liquid



Still not finding the right product?

Explore all of our products under 2-(Difluoromethoxy)ethan-1-amine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library