Skip to Content
Merck

Skip To

BDPH9BCDBB20

Bide Pharmatech

1-((Trifluoromethyl)sulfonyl)piperidine

Synonym(s):

1-((trifluoromethyl)sulfonyl)piperidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H10O2N1S1F3
CAS Number:
Molecular Weight:
217.21
MDL number:
Product BDPH9BCDBB20 is not currently sold in your country.


Inchi Code

1S/C6H10F3NO2S/c7-6(8,9)13(11,12)10-4-2-1-3-5-10/h1-5H2

Inchi Key

MBLHRHYLNYFCEV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library