Skip to Content
Merck

BDPH9BCDBC1D

Bide Pharmatech

3,9-Dibromo-5,11-dioctyl-5,11-dihydroindolo[3,2-b]carbazole

Synonym(s):

3,9-dibromo-5,11-dioctyl-5,11-dihydroindolo[3,2-b]carbazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C34H42N2Br2
CAS Number:
Molecular Weight:
638.53
MDL number:
Product BDPH9BCDBC1D is not currently sold in your country.


Inchi Code

1S/C34H42Br2N2/c1-3-5-7-9-11-13-19-37-31-21-25(35)15-17-27(31)29-24-34-30(23-33(29)37)28-18-16-26(36)22-32(28)38(34)20-14-12-10-8-6-4-2/h15-18,21-24H,3-14,19-20H2,1-2H3

Inchi Key

HFMSSMCMAMHDMB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library